Learn More
Sino Biological Recombinant Human ACE2 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10108H02H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 740) of human ACE2 precursor (NP_068576.1) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5505