Learn More
Medchemexpress LLC ACE2 Rat HEK293 H 10ug
SDP
Catalog No. 5000181695
Encompass_Preferred
ACE2 is an important carboxypeptidase in the renin-angiotensin system that complexly regulates cardiovascular homeostasis It specifically converts angiotensin I to angiotensin 1-9 and angiotensin II to angiotensin 1-7 exerting antihypertrophic and vasodilatory effects ACE2 Protein Rat (HEK293 His) is the recombinant rat-derived ACE2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81695 Supplier Medchemexpress LLC Supplier No. HYP7280910UG
ACE2 is an important carboxypeptidase in the renin-angiotensin system that complexly regulates cardiovascular homeostasis It specifically converts angiotensin I to angiotensin 1-9 and angiotensin II to angiotensin 1-7 exerting antihypertrophic and vasodilatory effects ACE2 Protein Rat (HEK293 His) is the recombinant rat-derived ACE2 protein expressed by HEK293 with C-His labeled tag