Learn More
Cayman Chemical ACE2 Reagent Concentrate 120uL
SDP
Catalog No. 502895216
Encompass_Preferred
A dual GLP-1R and GLP-2R agonist induces cAMP accumulation in HEK293 cells expressing human GLP-1R or GLP-2R (EC50s 190 and 210 pM respectively) reduces peak blood glucose levels in an oral glucose tolerance test and increases intestinal wet weight in mice at 6 nmol/kg decreases gastric emptying and increases intestinal weight and length in rats (ED50s 7.6 20.7 and 10.7 nmol/kg respectively) promotes body weight recovery decreases stool water content and increases intestinal villus height and intestinal length in a mouse model of short bowel and intestinal insufficiency induced by 40 ileocecal resection
Catalog No. 50-289-5216 Supplier Cayman Chemical Supplier No. 502072120 UL
Add to Cart
Add to Cart
A dual GLP-1R and GLP-2R agonist induces cAMP accumulation in HEK293 cells expressing human GLP-1R or GLP-2R (EC50s 190 and 210 pM respectively) reduces peak blood glucose levels in an oral glucose tolerance test and increases intestinal wet weight in mice at 6 nmol/kg decreases gastric emptying and increases intestinal weight and length in rats (ED50s 7.6 20.7 and 10.7 nmol/kg respectively) promotes body weight recovery decreases stool water content and increases intestinal villus height and intestinal length in a mouse model of short bowel and intestinal insufficiency induced by 40 ileocecal resection