Learn More
Creative Biomart Recombinant Human ACE2 Protein, Fc-tagged
Supplier:  Creative Biomart ACE2363
Encompass_Preferred
The CHO derived ACE2 Human recombinant protein contains theextracellular domain amino acids 19-740 fused to Fc tag at C-terminal. ACE2 Protein binds to SARS Coronavirus-2 [CoV-2019] Spike receptor bin
Catalog No. 50-208-4741