Learn More
Cayman Chemical ACE2humn Monclonal ntIb 100ug
SDP
Catalog No. 502991572
Encompass_Preferred
Immunogen: Recombinant human ACE2 • Host: Mouse • Species Reactivity: (+) Human • Applications: ELISA, FC, and WB
Catalog No. 50-299-1572 Supplier Cayman Chemical Supplier No. 30582100 UG
Immunogen: Recombinant human ACE2 • Host: Mouse • Species Reactivity: (+) Human • Applications: ELISA, FC, and WB