Learn More
Cayman Chemical ACE2humn Monclonal ntIb 100ug
SDP
Supplier:  Cayman Chemical 30584100 UG
Encompass_Preferred
Immunogen: Recombinant human ACE2 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Applications: ELISA and WB
Catalog No. 50-299-0907