Learn More
Cayman Chemical ACE2humn Monclonal ntIb 100ug
SDP
Catalog No. 502991878
Encompass_Preferred
Immunogen: Recombinant human ACE2 • Host: Mouse • Species Reactivity: (+) Human, mouse • Applications: ELISA, WB • MW: ~92 kDa
Catalog No. 50-299-1878 Supplier Cayman Chemical Supplier No. 31359100 UG
Immunogen: Recombinant human ACE2 • Host: Mouse • Species Reactivity: (+) Human, mouse • Applications: ELISA, WB • MW: ~92 kDa