Learn More
Enzo Life Sciences 5-Nitro-6,7-dichloro-1,4-dihydro-2,3-quinoxalinedione (25mg). CAS: 153504-81-5
SDP
Supplier:  Enzo Life Sciences ALX550228M025
Encompass_Preferred
Potent and selective NMDA receptor glycine site antagonist. Alternative name: ACEA-1021. Purity: ≥98% (TLC; elemental analysis). Appearance: Yellow powder. Solubility: Soluble in DMSO or dilute aqueous base. Long Term Storage: +4°C.
Catalog No. 50-200-9501