Learn More
Enzo Life Sciences Arachidonyl-2-chloroethylamide (50mg). CAS: 220556-69-4
SDP
Supplier:  Enzo Life Sciences BMLFA0320050
Encompass_Preferred
Non-pungent, potent and highly selective agonist of the cannabinoid CB1 receptor (Ki=1.4nM). Displays >1’400-fold selectivity over CB2 receptors (Ki=3.1µM). Active in vivo. Alternative name: ACEA, N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0814