Learn More
Cayman Chemical Acebilustat, Purity: ≥98%, CAS: 943764-99-6, 10 mg
SDP
Catalog No. 502950292
Encompass_Preferred
An inhibitor of LTA4 hydrolase; reduces the transmigration of PMNs from an epithelial monolayer to airway supernatant isolated from patients with cystic fibrosis at 1 µM
Catalog No. 50-295-0292 Supplier Cayman Chemical Supplier No. 3327010 MG
An inhibitor of LTA4 hydrolase; reduces the transmigration of PMNs from an epithelial monolayer to airway supernatant isolated from patients with cystic fibrosis at 1 µM