Learn More
Apexbio Technology LLC Acebutolol HCl 34381-68-5 1g
SDP
Catalog No. 5000091975
Encompass_Preferred
Acebutolol hydrochloride (CAS 34381-68-5) is a selective 1-adrenergic receptor antagonist By competitively inhibiting the binding of catecholamines to 1-adrenergic receptors acebutolol modulates sympathetic nervous system activity resulting in decreased heart rate and myocardial contractility This pharmacological profile underlies its utility in preclinical studies on cardiovascular regulation including models of hypertension angina pectoris and cardiac arrhythmias Acebutolol HCl is widely used in biomedical research to investigate -adrenergic signaling pathways and to evaluate therapeutic approaches targeting cardiac function and blood pressure control
Catalog No. 50-000-91975 Supplier Apexbio Technology LLC Supplier No. B13301000
Acebutolol hydrochloride (CAS 34381-68-5) is a selective 1-adrenergic receptor antagonist By competitively inhibiting the binding of catecholamines to 1-adrenergic receptors acebutolol modulates sympathetic nervous system activity resulting in decreased heart rate and myocardial contractility This pharmacological profile underlies its utility in preclinical studies on cardiovascular regulation including models of hypertension angina pectoris and cardiac arrhythmias Acebutolol HCl is widely used in biomedical research to investigate -adrenergic signaling pathways and to evaluate therapeutic approaches targeting cardiac function and blood pressure control