Learn More
Medchemexpress LLC HY-17497A 100mg Medchemexpress, Acebutolol (hydrochloride) CAS:34381-68-5 Purity:>98%
SDP
Catalog No. 502081968
Encompass_Preferred
Medchemexpress, HY-17497A 100mg Acebutolol (hydrochloride) CAS:34381-68-5 Acebutolol hydrochloride is a β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1968 Supplier Medchemexpress LLC Supplier No. HY17497A100MG
Medchemexpress, HY-17497A 100mg Acebutolol (hydrochloride) CAS:34381-68-5 Acebutolol hydrochloride is a β1 adrenergic receptor (β1AR) antagonist. Acebutolol hydrochloride is used in the treatment of hypertension, angina pectoris and cardiac arrhythmias. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.