Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acebutolol hydrochloride | 34381-68-5 | MFCD00078860 | 99.7% | 372.89 | 500 MG
SDP

Catalog No. 5000386860
Encompass_Preferred

Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist used to treat hypertension, angina pectoris, and cardiac arrhythmias.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture
  • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: 100 mg/mL (268.18 mM)
  • Solubility in H2O: ≥ 50 mg/mL (134.09 mM)

Catalog No. 50-003-86860 Supplier Medchemexpress LLC Supplier No. HY17497A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist used to treat hypertension, angina pectoris, and cardiac arrhythmias.

  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture
  • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: 100 mg/mL (268.18 mM)
  • Solubility in H2O: ≥ 50 mg/mL (134.09 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.