Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acebutolol hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111013978 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3978 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3978 Supplier MilliporeSigma Supelco Supplier No. A36691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acebutolol hydrochloride is a β-adrenergic receptor blocker. It is used in the treatment of hypertension, cardiac arrhythmias and angina pectoris.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acebutolol Hydrochloride
CAS 34381-68-5
Physical Form Neat
Linear Formula C18H28N2O4 · HCl
Technique Gas Chromatography
MDL Number MFCD00078860
Synonym N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide
Type Analytical Standard
Molecular Weight (g/mol) 372.89
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 372.89
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.