Learn More
TARGETMOL CHEMICALS INC ACECAINIDE HYDROCHLORIDE 25MG
SDP
Catalog No. 502580210
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acecainide hydrochloride (NAPA) is a major metabolite of PROCAINAMIDE. Its anti-arrhythmic action may cause cardiac toxicity in kidney failure. purity: 99%
Catalog No. 50-258-0210 Supplier TARGETMOL CHEMICALS INC Supplier No. T357525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Acecainide hydrochloride (NAPA) is a major metabolite of PROCAINAMIDE. Its anti-arrhythmic action may cause cardiac toxicity in kidney failure. purity: 99%