Learn More
Medchemexpress LLC Aceclidine hydrochloride | 6109-70-2 | 98.0% | 205.68 | 1 ML
SDP
Catalog No. 5000400652
Encompass_Preferred
Aceclidine hydrochloride is a modulator of the M3 muscarinic acetylcholine receptor and acts as an M1 receptor agonist with an EC50 of 40 μM. It functions as a cycloplegic agent, a surfactant, a tonicity adjustor, and can optionally serve as a viscosity enhancer and an antioxidant. This compound holds potential for research into various disorders, including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor.
  • M1 receptor agonist (EC50: 40 μM).
  • Functions as a cycloplegic agent.
  • Acts as a surfactant.
  • Serves as a tonicity adjustor.
  • Can optionally be a viscosity enhancer and an antioxidant.
  • Potential for research in disorders such as refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
Catalog No. 50-004-00652 Supplier Medchemexpress LLC Supplier No. HY32067A10MM/1ML
Aceclidine hydrochloride is a modulator of the M3 muscarinic acetylcholine receptor and acts as an M1 receptor agonist with an EC50 of 40 μM. It functions as a cycloplegic agent, a surfactant, a tonicity adjustor, and can optionally serve as a viscosity enhancer and an antioxidant. This compound holds potential for research into various disorders, including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor.
  • M1 receptor agonist (EC50: 40 μM).
  • Functions as a cycloplegic agent.
  • Acts as a surfactant.
  • Serves as a tonicity adjustor.
  • Can optionally be a viscosity enhancer and an antioxidant.
  • Potential for research in disorders such as refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.