Learn More
Medchemexpress LLC Aceclidine hydrochloride | 6109-70-2 | 98.0% | 205.68 | 500 MG
SDP
Catalog No. 5000400440
Encompass_Preferred
Aceclidine hydrochloride is identified as a modulator of the M3 muscarinic acetylcholine receptor and an M1 receptor agonist, with an EC50 of 40 μM. It functions as a cycloplegic agent, a surfactant, a tonicity adjustor, and can optionally act as a viscosity enhancer and an antioxidant. This product holds potential for research into various disorders, including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor and an M1 receptor agonist (EC50: 40 μM).
  • Acts as a cycloplegic agent.
  • Functions as a surfactant.
  • Serves as a tonicity adjustor.
  • Can be used as a viscosity enhancer.
  • Possesses antioxidant properties.
  • Potential for research in eye-related conditions and disorders such as xerostomia and Sjogren's syndrome.
Catalog No. 50-004-00440 Supplier Medchemexpress LLC Supplier No. HY32067A500MG
Aceclidine hydrochloride is identified as a modulator of the M3 muscarinic acetylcholine receptor and an M1 receptor agonist, with an EC50 of 40 μM. It functions as a cycloplegic agent, a surfactant, a tonicity adjustor, and can optionally act as a viscosity enhancer and an antioxidant. This product holds potential for research into various disorders, including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor and an M1 receptor agonist (EC50: 40 μM).
  • Acts as a cycloplegic agent.
  • Functions as a surfactant.
  • Serves as a tonicity adjustor.
  • Can be used as a viscosity enhancer.
  • Possesses antioxidant properties.
  • Potential for research in eye-related conditions and disorders such as xerostomia and Sjogren's syndrome.