Learn More
Medchemexpress LLC Aceclidine hydrochloride | 6109-70-2 | MFCD00468105 | 98.0% | 205.68 | 1 G
SDP
Catalog No. 5000401970
Encompass_Preferred
Aceclidine (Quinuclidin-3-yl acetate) hydrochloride is a modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM). It is also a cycloplegic agent, a surfactant, a tonicity adjustor, and optionally a viscosity enhancer and an antioxidant. It has potential for research into various disorders including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM)
  • Cycloplegic agent
  • Surfactant
  • Tonicity adjustor
  • Optionally a viscosity enhancer and an antioxidant
  • Potential for research of disorders such as refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia
Catalog No. 50-004-01970 Supplier Medchemexpress LLC Supplier No. HY32067A1G
Aceclidine (Quinuclidin-3-yl acetate) hydrochloride is a modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM). It is also a cycloplegic agent, a surfactant, a tonicity adjustor, and optionally a viscosity enhancer and an antioxidant. It has potential for research into various disorders including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM)
  • Cycloplegic agent
  • Surfactant
  • Tonicity adjustor
  • Optionally a viscosity enhancer and an antioxidant
  • Potential for research of disorders such as refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia