Learn More
Medchemexpress LLC Aceclidine hydrochloride (quinuclidin-3-yl acetate hydrochloride) | 6109-70-2 | 98.0% | 205.68 | 100 MG
SDP
Catalog No. 5000401297
Encompass_Preferred
Aceclidine hydrochloride is a modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM). It acts as a cycloplegic agent, surfactant, tonicity adjustor, viscosity enhancer, and antioxidant. It has potential for research into refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of muscarinic acetylcholine receptor.
  • Agonist of muscarinic acetylcholine receptor.
  • Cycloplegic agent.
  • Surfactant.
  • Tonicity adjustor.
  • Viscosity enhancer.
  • Antioxidant.
  • Potential for research of refractive errors of the eye.
  • Potential for research of xerostomia.
  • Potential for research of Sjogren's syndrome.
  • Potential for research of glaucoma.
  • Potential for research of conjunctivitis.
  • Potential for research of lacrimal gland disease.
  • Potential for research of esotropia.
Catalog No. 50-004-01297 Supplier Medchemexpress LLC Supplier No. HY32067A100MG
Aceclidine hydrochloride is a modulator of M3 muscarinic acetylcholine receptor and a M1 receptor agonist (EC50: 40 μM). It acts as a cycloplegic agent, surfactant, tonicity adjustor, viscosity enhancer, and antioxidant. It has potential for research into refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulator of muscarinic acetylcholine receptor.
  • Agonist of muscarinic acetylcholine receptor.
  • Cycloplegic agent.
  • Surfactant.
  • Tonicity adjustor.
  • Viscosity enhancer.
  • Antioxidant.
  • Potential for research of refractive errors of the eye.
  • Potential for research of xerostomia.
  • Potential for research of Sjogren's syndrome.
  • Potential for research of glaucoma.
  • Potential for research of conjunctivitis.
  • Potential for research of lacrimal gland disease.
  • Potential for research of esotropia.