Learn More
Cayman Chemical Aceclidine (hydrochloride), Purity: ≥95%, CAS: 6109-70-2, 100 mg
SDP
Catalog No. 502966540
Encompass_Preferred
An agonist of muscarinic receptors (EC50 = 1.8-17 µM for human M2)
Catalog No. 50-296-6540 Supplier Cayman Chemical Supplier No. 17910100 MG
An agonist of muscarinic receptors (EC50 = 1.8-17 µM for human M2)