Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aceclofenac | 89796-99-6 | 99.8% | 5 G
SDP

Catalog No. 5000408735
Encompass_Preferred

Aceclofenac is an orally active nonsteroidal anti-inflammatory agent (NSAID) with analgesic and anti-inflammatory properties. It is utilized in research for conditions such as osteoarthritis, ankylosing spondylitis, and rheumatoid arthritis.

  • Orally active nonsteroidal anti-inflammatory agent (NSAID)
  • Possesses analgesic and anti-inflammatory properties
  • Molecular formula: C16H13Cl2NO4
  • Molecular weight: 354.18
  • Appearance: white to off-white solid
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-08735 Supplier Medchemexpress LLC Supplier No. HYB06345G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aceclofenac is an orally active nonsteroidal anti-inflammatory agent (NSAID) with analgesic and anti-inflammatory properties. It is utilized in research for conditions such as osteoarthritis, ankylosing spondylitis, and rheumatoid arthritis.

  • Orally active nonsteroidal anti-inflammatory agent (NSAID)
  • Possesses analgesic and anti-inflammatory properties
  • Molecular formula: C16H13Cl2NO4
  • Molecular weight: 354.18
  • Appearance: white to off-white solid
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.