Learn More
Cayman Chemical Aceclofenac ethyl ester, Purity: ≥98%, CAS: 139272-67-6, 10 mg
SDP
Catalog No. 502985884
Encompass_Preferred
A potential impurity in commercial preparations of aceclofenac
Catalog No. 50-298-5884 Supplier Cayman Chemical Supplier No. 2211510 MG
A potential impurity in commercial preparations of aceclofenac