Learn More
Cayman Chemical Aceclofenac ethyl ester 50mg
SDP
Catalog No. 502986625
Encompass_Preferred
A potential impurity in commercial preparations of aceclofenac
Catalog No. 50-298-6625 Supplier Cayman Chemical Supplier No. 2211550 MG
A potential impurity in commercial preparations of aceclofenac