Learn More
Cayman Chemical Aceclofenac methyl ester 10mg
SDP
Catalog No. 502987232
Encompass_Preferred
A potential impurity in commercial preparations of aceclofenac; has been used as a precursor in the synthesis of aceclofenac
Catalog No. 50-298-7232 Supplier Cayman Chemical Supplier No. 2212110 MG
A potential impurity in commercial preparations of aceclofenac; has been used as a precursor in the synthesis of aceclofenac