Learn More
Cayman Chemical Aceclofenac methyl ester, Purity: ≥98%, CAS: 139272-66-5, 50 mg
SDP
Catalog No. 502987229
Encompass_Preferred
A potential impurity in commercial preparations of aceclofenac; has been used as a precursor in the synthesis of aceclofenac
Catalog No. 50-298-7229 Supplier Cayman Chemical Supplier No. 2212150 MG
A potential impurity in commercial preparations of aceclofenac; has been used as a precursor in the synthesis of aceclofenac