Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aceclofenac 98.0+%, TCI America™
SDP

Catalog No. A24841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24841G 1 g
A24845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24841G Supplier TCI America Supplier No. A24841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89796-99-6
Molecular Formula C16H13Cl2NO4
Molecular Weight (g/mol) 354.183
MDL Number MFCD00864296
InChI Key MNIPYSSQXLZQLJ-UHFFFAOYSA-N
Synonym aceclofenac, aceclofenaco, aceclofenacum, preservex, airtal, falcol, gerbin, aceclofar, aceclofenacum latin, aceclofenaco spanish
PubChem CID 71771
ChEBI CHEBI:31159
IUPAC Name 2-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxyacetic acid
SMILES C1=CC=C(C(=C1)CC(=O)OCC(=O)O)NC2=C(C=CC=C2Cl)Cl

Specifications

Melting Point 152°C
Color Yellow
UN Number 2811
Quantity 1 g
Formula Weight 354.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Aceclofenac
RTECSNumber CY1577900
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.