Learn More
Medchemexpress LLC Acedoben | 556-08-1 | 99.9% | 25 G
SDP
Catalog No. 5000413739
Encompass_Preferred
Acedoben is a biochemical agent that can form a fast self-assembled coordination complex with iron ions. The Fe-Ace coordination complex can serve as a carrier of tumor antigens and enhance antigen-specific anti-tumor immunity. This product is for research use only.
  • Forms coordination complex with iron ions
  • Enhances antigen-specific anti-tumor immunity
  • Suitable for laboratory research applications
Catalog No. 50-004-13739 Supplier Medchemexpress LLC Supplier No. 25GHYB1250
Acedoben is a biochemical agent that can form a fast self-assembled coordination complex with iron ions. The Fe-Ace coordination complex can serve as a carrier of tumor antigens and enhance antigen-specific anti-tumor immunity. This product is for research use only.
  • Forms coordination complex with iron ions
  • Enhances antigen-specific anti-tumor immunity
  • Suitable for laboratory research applications