Learn More
Medchemexpress LLC Acedoben | 556-08-1 | 99.9% | 50 G
SDP
Catalog No. 5000412815
Encompass_Preferred
Acedoben is a biochemical agent that can form a fast self-assembled coordination complex with iron ions. This Fe-Ace coordination complex acts as a carrier of tumor antigens and enhances antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
  • Functions as a biochemical agent.
  • Constructs a fast self-assembled coordination complex with iron ions.
  • Serves as a carrier of tumor antigens.
  • Enhances antigen-specific anti-tumor immunity.
  • Possesses inherent adjuvant properties.
  • Available in solid form.
  • Soluble in DMSO.
Catalog No. 50-004-12815 Supplier Medchemexpress LLC Supplier No. HYB125050G
Add to Cart
Add to Cart
Acedoben is a biochemical agent that can form a fast self-assembled coordination complex with iron ions. This Fe-Ace coordination complex acts as a carrier of tumor antigens and enhances antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
  • Functions as a biochemical agent.
  • Constructs a fast self-assembled coordination complex with iron ions.
  • Serves as a carrier of tumor antigens.
  • Enhances antigen-specific anti-tumor immunity.
  • Possesses inherent adjuvant properties.
  • Available in solid form.
  • Soluble in DMSO.