Learn More
Medchemexpress LLC Acedoben (Standard) | 556-08-1 | 250 MG
SDP
Catalog No. 5000414122
Encompass_Preferred
Acedoben (Standard) is the analytical standard of Acedoben, intended for research and analytical applications. It is a biochemical agent. Acedoben and iron ions can construct a fast self-assembled coordination complex. The Fe-Ace coordination complex can not only serve as a carrier of tumor antigens, but also enhance antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
  • Analytical standard of Acedoben.
  • Intended for research and analytical applications.
  • Biochemical agent.
  • Forms a fast, self-assembled coordination complex with iron ions.
  • Fe-Ace coordination complex can serve as a carrier of tumor antigens.
  • Enhances antigen-specific anti-tumor immunity.
Catalog No. 50-004-14122 Supplier Medchemexpress LLC Supplier No. 250MGHYB1250R
Acedoben (Standard) is the analytical standard of Acedoben, intended for research and analytical applications. It is a biochemical agent. Acedoben and iron ions can construct a fast self-assembled coordination complex. The Fe-Ace coordination complex can not only serve as a carrier of tumor antigens, but also enhance antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
  • Analytical standard of Acedoben.
  • Intended for research and analytical applications.
  • Biochemical agent.
  • Forms a fast, self-assembled coordination complex with iron ions.
  • Fe-Ace coordination complex can serve as a carrier of tumor antigens.
  • Enhances antigen-specific anti-tumor immunity.