Learn More
Medchemexpress LLC Acefylline | 652-37-9 | 99.5% | 1 ML
SDP
Catalog No. 5000411954
Encompass_Preferred
Acefylline (Theophyllineacetic acid) is a xanthine derivative that acts as an adenosine receptor antagonist and a peptidylarginine deiminase (PAD) activator. It also functions as a bronchodilator and cardiac stimulant by inhibiting rat lung cAMP phosphodiesterase isoenzymes, making it suitable for asthma research.
  • Xanthine derivative
  • Adenosine receptor antagonist
  • Peptidylarginine deiminase (PAD) activator
  • Bronchodilator and cardiac stimulant
  • Inhibits rat lung cAMP phosphodiesterase isoenzymes
  • Applicable in asthma research
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Structure classification: Ketones, aldehydes, acids
Catalog No. 50-004-11954 Supplier Medchemexpress LLC Supplier No. HYB150510MM/1ML
Acefylline (Theophyllineacetic acid) is a xanthine derivative that acts as an adenosine receptor antagonist and a peptidylarginine deiminase (PAD) activator. It also functions as a bronchodilator and cardiac stimulant by inhibiting rat lung cAMP phosphodiesterase isoenzymes, making it suitable for asthma research.
  • Xanthine derivative
  • Adenosine receptor antagonist
  • Peptidylarginine deiminase (PAD) activator
  • Bronchodilator and cardiac stimulant
  • Inhibits rat lung cAMP phosphodiesterase isoenzymes
  • Applicable in asthma research
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite
  • Structure classification: Ketones, aldehydes, acids