Learn More
Medchemexpress LLC Acelarin (NUC-1031) | 840506-29-8 | 99.1% | 580.47 g/mol | C25H27F2N4O8P | 25 MG
SDP
Catalog No. 5000211574
Encompass_Preferred
Acelarin (NUC-1031) is a ProTide transformation of the nucleoside analogue gemcitabine developed for research use. It improves intracellular delivery and metabolic stability, demonstrating potent in vitro activity and tumor-growth reduction in preclinical models. Supplied as a solid and as a DMSO solution, the material includes specification and storage guidance for research applications.
  • Enhances cellular uptake compared with parent nucleoside.
  • Resists deamination and bypasses kinase dependence.
  • Potent inhibition of DNA synthesis in vitro.
  • Available as solid quantities and DMSO solution for flexibility.
  • Long-term stable under recommended storage conditions.
Catalog No. 50-002-11574 Supplier Medchemexpress LLC Supplier No. HY10088525MG
Acelarin (NUC-1031) is a ProTide transformation of the nucleoside analogue gemcitabine developed for research use. It improves intracellular delivery and metabolic stability, demonstrating potent in vitro activity and tumor-growth reduction in preclinical models. Supplied as a solid and as a DMSO solution, the material includes specification and storage guidance for research applications.
  • Enhances cellular uptake compared with parent nucleoside.
  • Resists deamination and bypasses kinase dependence.
  • Potent inhibition of DNA synthesis in vitro.
  • Available as solid quantities and DMSO solution for flexibility.
  • Long-term stable under recommended storage conditions.