Learn More
MARIAN MEDICAL INC ACElok Dressing, For ACE Stopper, 10 Per Box
Catalog No. NC3958259
Encompass_Preferred
ACE Dressing. Stoma Dressing. Marian Medical Inc. SD03. External fixation and cushioning dressing designed for use with ACE stoppers. Allows access to stoma site during washout procedures. Used to support secure positioning of ACE stoppers. For stoma care applications. 10 per box.
Catalog No. NC3958259 Supplier MARIAN MEDICAL INC Supplier No. SD03
ACE Dressing. Stoma Dressing. Marian Medical Inc. SD03. External fixation and cushioning dressing designed for use with ACE stoppers. Allows access to stoma site during washout procedures. Used to support secure positioning of ACE stoppers. For stoma care applications. 10 per box.