Learn More
Apexbio Technology LLC Acemetacin 53164-05-9 1g
SDP
Catalog No. 502240805
Encompass_Preferred
Acemetacin (CAS 53164-05-9) is a non-steroidal anti-inflammatory compound structurally classified as a glycolic acid ester derivative of indometacin It exerts its pharmacological effects by inhibiting cyclooxygenase (COX) enzymes thereby suppressing prostaglandin synthesis involved in inflammation and pain signaling pathways Acemetacin is commonly employed in biomedical research to investigate COX-mediated processes and the modulation of inflammatory responses as well as to explore pathways related to non-steroidal anti-inflammatory drug (NSAID) pharmacodynamics and safety profiles
Catalog No. 50-224-0805 Supplier Apexbio Technology LLC Supplier No. B14401000
Acemetacin (CAS 53164-05-9) is a non-steroidal anti-inflammatory compound structurally classified as a glycolic acid ester derivative of indometacin It exerts its pharmacological effects by inhibiting cyclooxygenase (COX) enzymes thereby suppressing prostaglandin synthesis involved in inflammation and pain signaling pathways Acemetacin is commonly employed in biomedical research to investigate COX-mediated processes and the modulation of inflammatory responses as well as to explore pathways related to non-steroidal anti-inflammatory drug (NSAID) pharmacodynamics and safety profiles