Learn More
Medchemexpress LLC Acemetacin | 53164-05-9 | 99.95% | 1 ML
SDP
Catalog No. 5000409443
Encompass_Preferred
Acemetacin (TVX 1322) is a non-steroidal anti-inflammatory drug and a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor. For research use only. It is used for the treatment of osteoarthritis, rheumatoid arthritis, lower back pain, and relieving post-operative pain. Acemetacin acts as a prodrug; in the body, it is metabolized to indometacin, which then acts as an inhibitor of cyclooxygenase, producing the anti-inflammatory effects. An advantage of Acemetacin is that it reduces gastric damage when compared to indometacin.
  • Non-steroidal anti-inflammatory drug
  • Glycolic acid ester of indometacin
  • Cyclooxygenase inhibitor
  • Prodrug metabolized to indometacin in the body
  • Reduces gastric damage compared to indometacin
Catalog No. 50-004-09443 Supplier Medchemexpress LLC Supplier No. HYB048210MM/1ML
Acemetacin (TVX 1322) is a non-steroidal anti-inflammatory drug and a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor. For research use only. It is used for the treatment of osteoarthritis, rheumatoid arthritis, lower back pain, and relieving post-operative pain. Acemetacin acts as a prodrug; in the body, it is metabolized to indometacin, which then acts as an inhibitor of cyclooxygenase, producing the anti-inflammatory effects. An advantage of Acemetacin is that it reduces gastric damage when compared to indometacin.
  • Non-steroidal anti-inflammatory drug
  • Glycolic acid ester of indometacin
  • Cyclooxygenase inhibitor
  • Prodrug metabolized to indometacin in the body
  • Reduces gastric damage compared to indometacin