Learn More
Medchemexpress LLC Acemetacin (Standard) | 53164-05-9 | MFCD00151473 | >98.0% | 1 G
SDP
Catalog No. 5000406414
Encompass_Preferred
Acemetacin (Standard) is an analytical standard intended for research and analytical applications. Known also as TVX 1322, this compound is a non-steroidal anti-inflammatory drug (NSAID) and a glycolic acid ester of indometacin. It functions primarily as a cyclooxygenase inhibitor.
  • Analytical standard for research
  • Intended for analytical applications
  • Functions as a non-steroidal anti-inflammatory drug (NSAID)
  • Glycolic acid ester of indometacin
  • Inhibits cyclooxygenase (COX)
Catalog No. 50-004-06414 Supplier Medchemexpress LLC Supplier No. HYB04821G
Acemetacin (Standard) is an analytical standard intended for research and analytical applications. Known also as TVX 1322, this compound is a non-steroidal anti-inflammatory drug (NSAID) and a glycolic acid ester of indometacin. It functions primarily as a cyclooxygenase inhibitor.
  • Analytical standard for research
  • Intended for analytical applications
  • Functions as a non-steroidal anti-inflammatory drug (NSAID)
  • Glycolic acid ester of indometacin
  • Inhibits cyclooxygenase (COX)