Learn More
Cayman Chemical Acenaphthene-d10, Purity: ≥99% deuterated forms (d1-d10), CAS: 15067-26-2, 1 g
SDP
Catalog No. 502918956
Encompass_Preferred
An internal standard for the quantification of acenaphthene by GC- or LC-MS
Catalog No. 50-291-8956 Supplier Cayman Chemical Supplier No. 389681 G
An internal standard for the quantification of acenaphthene by GC- or LC-MS