Learn More
Medchemexpress LLC Acenaphthylene | 208-96-8 | 99.4% | 152.20 | 1 G
SDP
Catalog No. 5000454526
Encompass_Preferred
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are naturally derived from coal and tar deposits and are produced by the incomplete combustion of organic matter.
  • Polycyclic aromatic hydrocarbon (PAH)
  • Naturally derived from coal and tar deposits
  • Produced by incomplete combustion of organic matter
  • Solid appearance, light yellow to yellow color
Catalog No. 50-004-54526 Supplier Medchemexpress LLC Supplier No. HYW0135701G
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are naturally derived from coal and tar deposits and are produced by the incomplete combustion of organic matter.
  • Polycyclic aromatic hydrocarbon (PAH)
  • Naturally derived from coal and tar deposits
  • Produced by incomplete combustion of organic matter
  • Solid appearance, light yellow to yellow color