Learn More
Medchemexpress LLC Acenaphthylene | 208-96-8 | 99.4% | 152.20 | 25 G
SDP
Catalog No. 5000366692
Encompass_Preferred
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH) and an analytical standard intended for research and analytical applications. PAHs are naturally derived from coal and tar deposits, and produced by incomplete combustion of organic matter.
  • Solid appearance, light yellow to yellow in color
  • Polycyclic aromatic hydrocarbon (PAH)
  • Intended for research and analytical applications
  • Good solubility in DMSO
Catalog No. 50-003-66692 Supplier Medchemexpress LLC Supplier No. 25GHYW013570
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH) and an analytical standard intended for research and analytical applications. PAHs are naturally derived from coal and tar deposits, and produced by incomplete combustion of organic matter.
  • Solid appearance, light yellow to yellow in color
  • Polycyclic aromatic hydrocarbon (PAH)
  • Intended for research and analytical applications
  • Good solubility in DMSO