Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acenaphthylene | 208-96-8 | 99.4% | 152.20 | 5 G
SDP

Catalog No. 5000454466
Encompass_Preferred

Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are derived naturally from coal and tar deposits, and produced by incomplete combustion of organic matter.

  • Molecular weight: 152.20
  • Formula: C12H8
  • CAS no.: 208-96-8
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Others
  • Initial source: Plants other families

Catalog No. 50-004-54466 Supplier Medchemexpress LLC Supplier No. HYW0135705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are derived naturally from coal and tar deposits, and produced by incomplete combustion of organic matter.

  • Molecular weight: 152.20
  • Formula: C12H8
  • CAS no.: 208-96-8
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Others
  • Initial source: Plants other families

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.