Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acenaphthylene | 208-96-8 | 99.4% | 152.20 | 50 G
SDP

Catalog No. 5000454710
Encompass_Preferred

Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH) naturally derived from coal and tar deposits and produced by incomplete combustion of organic matter. This product is intended for research and analytical applications.

  • Molecular formula: C12H8
  • Appearance: Solid
  • Color: Light yellow to yellow
  • CAS number: 208-96-8
  • Molecular weight: 152.20
  • Purity: 99.44%
  • Shipping condition for continental US: Room temperature
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Not validated for medical applications; for research use only

Catalog No. 50-004-54710 Supplier Medchemexpress LLC Supplier No. HYW01357050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH) naturally derived from coal and tar deposits and produced by incomplete combustion of organic matter. This product is intended for research and analytical applications.

  • Molecular formula: C12H8
  • Appearance: Solid
  • Color: Light yellow to yellow
  • CAS number: 208-96-8
  • Molecular weight: 152.20
  • Purity: 99.44%
  • Shipping condition for continental US: Room temperature
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Not validated for medical applications; for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.