Learn More
Medchemexpress LLC Acenaphthylene | 208-96-8 | MFCD00003806 | 99.44% | 152.20 | 100 G
SDP
Catalog No. 5000454482
Encompass_Preferred
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are naturally derived from coal and tar deposits and produced by incomplete combustion of organic matter.
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Others
  • Initial source: Plants other families
Catalog No. 50-004-54482 Supplier Medchemexpress LLC Supplier No. HYW013570100G
Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are naturally derived from coal and tar deposits and produced by incomplete combustion of organic matter.
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Others
  • Initial source: Plants other families