Learn More
Medchemexpress LLC Acenaphthylene-d8 | 93951-97-4 | 99.5% | 160.24 | 25 MG
SDP
Catalog No. 5000452050
Encompass_Preferred
Acenaphthylene-d8 is the deuterium labeled Acenaphthylene, a polycyclic aromatic hydrocarbon (PAH). PAHs are derived naturally from coal and tar deposits, and produced by incomplete combustion of organic matter. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for research applications.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52050 Supplier Medchemexpress LLC Supplier No. 25MGHYW013570S
Acenaphthylene-d8 is the deuterium labeled Acenaphthylene, a polycyclic aromatic hydrocarbon (PAH). PAHs are derived naturally from coal and tar deposits, and produced by incomplete combustion of organic matter. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for research applications.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS