Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acenaphthylene, MilliporeSigma™ Supelco™

Catalog No. 111019542 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9542 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9542 Supplier MilliporeSigma Supelco Supplier No. 48566
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 208-96-8
Density 0.899 g/mL (at 25°C)
Flash Point 122°C
Physical Form Neat
Linear Formula C12H8
MDL Number MFCD00003806
Melting Point 78°C to 82°C
Boiling Point 280°C
Type Organic Pollutant Standard
Quantity 100 mg
Formula Weight 152.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.