Learn More
Cayman Chemical Acenocoumarol-d4 500ug
SDP
Catalog No. 502913567
Encompass_Preferred
An internal standard for the quantification of acenocoumarol by GC- or LC-MS
Catalog No. 50-291-3567 Supplier Cayman Chemical Supplier No. 35477500 UG
An internal standard for the quantification of acenocoumarol by GC- or LC-MS