Learn More
Cayman Chemical Acephate-d3 10mg
SDP
Catalog No. 502919209
Encompass_Preferred
An internal standard for the quantification of acephate by GC- or LC-MS
Catalog No. 50-291-9209 Supplier Cayman Chemical Supplier No. 3957910 MG
An internal standard for the quantification of acephate by GC- or LC-MS