Learn More
Cayman Chemical Acephate-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2140327-70-2, 1 mg
SDP
Catalog No. 502915931
Encompass_Preferred
An internal standard for the quantification of acephate by GC- or LC-MS
Catalog No. 50-291-5931 Supplier Cayman Chemical Supplier No. 395791 MG
An internal standard for the quantification of acephate by GC- or LC-MS