Learn More
Med Vet International Acepromazine Tablets, 25mg, 500ct., Generic Of Promace
Catalog No. 502828700
Encompass_Preferred
Acepromazine is a generic form of PromAce. It can act as an aid in tranquilization and as a preanesthetic agent in dogs. Acepromazine tablets may be used as an aid in controlling intractable animal during examination, treatment, grooming, x-ray, and minor surgical procedures.
Catalog No. 50-282-8700 Supplier Med Vet International Supplier No. RXACE25500
Acepromazine is a generic form of PromAce. It can act as an aid in tranquilization and as a preanesthetic agent in dogs. Acepromazine tablets may be used as an aid in controlling intractable animal during examination, treatment, grooming, x-ray, and minor surgical procedures.