Learn More
Cayman Chemical Acepromazine (maleate), Purity: ≥98%, CAS: 3598-37-6, 50 mg
SDP
Catalog No. 502939106
Encompass_Preferred
A phenothiazine antipsychotic and a dopamine receptor antagonist; binds to muscarinic receptors in rat brain (IC50 = 350 nM) and inhibits acetylcholine-induceed contractions of guinea pig ileum (IC50 = 840 nM); inhibits morphine-induced vomiting in dogs; inhibits development of a conditioned avoidance response, unconditioned escape response, and catalepsy in rats (ED50s = 0.98, 3.1, and 17 mg/kg, respectively)
Catalog No. 50-293-9106 Supplier Cayman Chemical Supplier No. 2782150 MG
A phenothiazine antipsychotic and a dopamine receptor antagonist; binds to muscarinic receptors in rat brain (IC50 = 350 nM) and inhibits acetylcholine-induceed contractions of guinea pig ileum (IC50 = 840 nM); inhibits morphine-induced vomiting in dogs; inhibits development of a conditioned avoidance response, unconditioned escape response, and catalepsy in rats (ED50s = 0.98, 3.1, and 17 mg/kg, respectively)