Learn More
Cayman Chemical AcequInocyl-d25 500ug
SDP
Supplier:  Cayman Chemical 39589500 UG
Encompass_Preferred
An internal standard for the quantification of acequinocyl by GC- or LC-MS
Catalog No. 50-291-6890