Learn More
Aviva Systems Biology Corp ACER1 Antibody - C-terminal region (ARP60813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACER1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ACER1. Alternate names: ASAH3, MGC138327, MGC138329, ALKCDase1ApplicationWB. Species reactivity: Human
Catalog No. 50-287-174